Anti-WDR5 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB22512-100EA 790 EUR
ABCAAB22512-100
Anti-WDR5 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: WDR5 (WD repeat domain 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR