Anti-CD200 / OX2 Mouse monoclonal antibody [clone: MRC OX-2]

Proveedor: Abcam
ABCAAB22353-100EA 684 EUR
ABCAAB22353-100
Anti-CD200 / OX2 Mouse monoclonal antibody [clone: MRC OX-2]
Type: Primary
Antigen: CD200 / OX2
Clonality: monoclonal
Clone: MRC OX-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR