Anti-CD74 Mouse monoclonal antibody [clone: MRC OX3]

ABCAAB22348-100EA 731 EUR
ABCAAB22348-100
Anti-CD74 Mouse monoclonal antibody [clone: MRC OX3]
Type: Primary
Antigen: CD74 (CD74 molecule, major histocompatibility complex, class II invariant chain)
Clonality: monoclonal
Clone: MRC OX3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR