Anti-ATF6 Rabbit polyclonal antibody

ABCAAB22341-100EA 0 EUR
ABCAAB22341-100
Anti-ATF6 Rabbit polyclonal antibody
Type: Primary
Antigen: ATF6 (activating transcription factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR