Anti-CAMK2B Rabbit polyclonal antibody

ABCAAB22183-50EA 588 EUR
ABCAAB22183-50
Anti-CAMK2B Rabbit polyclonal antibody
Type: Primary
Antigen: CAMK2B (calcium/calmodulin-dependent protein kinase II beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR