Anti-NFKBIA Rabbit polyclonal antibody

ABCAAB22071-100EA 588 EUR
ABCAAB22071-100
Anti-NFKBIA Rabbit polyclonal antibody
Type: Primary
Antigen: NFKBIA (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR