Anti-IKB zeta Rabbit polyclonal antibody

ABCAAB22065-50EA 588 EUR
ABCAAB22065-50
Anti-IKB zeta Rabbit polyclonal antibody
Type: Primary
Antigen: IKB zeta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR