Anti-TLR6 Mouse monoclonal antibody [clone: 86B1153.2]

ABCAAB22046-100EA 588 EUR
ABCAAB22046-100
Anti-TLR6 Mouse monoclonal antibody [clone: 86B1153.2]
Type: Primary
Antigen: TLR6 (toll-like receptor 6)
Clonality: monoclonal
Clone: 86B1153.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR