Anti-TP73 Mouse monoclonal antibody [clone: 5B1288]

ABCAAB22045-100EA 634 EUR
ABCAAB22045-100
Anti-TP73 Mouse monoclonal antibody [clone: 5B1288]
Type: Primary
Antigen: TP73 (tumor protein p73)
Clonality: monoclonal
Clone: 5B1288
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR