Anti-NFkB Inducing Kinase NIK Rabbit polyclonal antibody

ABCAAB22042-100EA 651 EUR
ABCAAB22042-100
Anti-NFkB Inducing Kinase NIK Rabbit polyclonal antibody
Type: Primary
Antigen: NFkB Inducing Kinase NIK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR