Anti-gamma Adaptin Rabbit polyclonal antibody

ABCAAB21980-100EA 661 EUR
ABCAAB21980-100
Anti-gamma Adaptin Rabbit polyclonal antibody
Type: Primary
Antigen: gamma Adaptin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR