Anti-ST3GAL1 Mouse polyclonal antibody

ABCAAB21948-100EA 588 EUR
ABCAAB21948-100
Anti-ST3GAL1 Mouse polyclonal antibody
Type: Primary
Antigen: ST3GAL1 (ST3 beta-galactoside alpha-2,3-sialyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR