Anti-CCR5 Rabbit polyclonal antibody

ABCAAB21653-100EA 577 EUR
ABCAAB21653-100
Anti-CCR5 Rabbit polyclonal antibody
Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR