Anti-LSP1 Rabbit polyclonal antibody

ABCAAB21636-50EA 604 EUR
ABCAAB21636-50
Anti-LSP1 Rabbit polyclonal antibody
Type: Primary
Antigen: LSP1 (lymphocyte-specific protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR