Anti-REST / NRSF Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB21635-100EA 790 EUR
ABCAAB21635-100
Anti-REST / NRSF Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: REST / NRSF
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR