Anti-Tropoelastin Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB21600-100EA 770 EUR
ABCAAB21600-100
Anti-Tropoelastin Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Tropoelastin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR