Anti-Neutrophil Elastase Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB21595-250EA 779 EUR
ABCAAB21595-250
Anti-Neutrophil Elastase Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Neutrophil Elastase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR