Anti-IL3RA Mouse Monoclonal Antibody [clone: 6H6]

Proveedor: Abcam
ABCAAB21562-100EA 495 EUR
ABCAAB21562-100
Anti-IL3RA Mouse Monoclonal Antibody [clone: 6H6]
Anticuerpos
Type: Primary
Antigen: IL3RA (interleukin 3 receptor, alpha (low affinity))
Clonality: monoclonal
Clone: 6H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR