Anti-DDDDK tag Rabbit polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB21536-100EA 0 EUR
ABCAAB21536-100
Anti-DDDDK tag Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: DDDDK tag (DDDDK tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR