Anti-Avian Influenza A Hemagglutinin Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB21292-100EA 739 EUR
ABCAAB21292-100
Anti-Avian Influenza A Hemagglutinin Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Avian Influenza A Hemagglutinin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR