Anti-PKNOX1.1 Chicken polyclonal antibody

ABCAAB21279-100EA 604 EUR
ABCAAB21279-100
Anti-PKNOX1.1 Chicken polyclonal antibody
Type: Primary
Antigen: PKNOX1.1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR