Anti-AGAP2 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB21271-100EA 744 EUR
ABCAAB21271-100
Anti-AGAP2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: AGAP2 (ArfGAP with GTPase domain, ankyrin repeat and PH domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR