Anti-XAGE1A Goat polyclonal antibody

ABCAAB21198-100EA 401 EUR
ABCAAB21198-100
Anti-XAGE1A Goat polyclonal antibody
Type: Primary
Antigen: XAGE1A (X antigen family, member 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR