Anti-ERC1 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB21192-100EA 745 EUR
ABCAAB21192-100
Anti-ERC1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ERC1 (ELKS/RAB6-interacting/CAST family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR