Anti-Adenovirus hexon protein Goat polyclonal antibody Biotin

ABCAAB21116-100EA 588 EUR
ABCAAB21116-100
Anti-Adenovirus hexon protein Goat polyclonal antibody Biotin
Type: Primary
Antigen: Adenovirus hexon protein
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR