Anti-Valproic Acid Mouse monoclonal antibody [clone: 804]

ABCAAB21000-500EA 588 EUR
ABCAAB21000-500
Anti-Valproic Acid Mouse monoclonal antibody [clone: 804]
Type: Primary
Antigen: Valproic Acid
Clonality: monoclonal
Clone: 804
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR