Anti-IL4 Rat monoclonal antibody [clone: I4.F5]

ABCAAB20989-100EA 604 EUR
ABCAAB20989-100
Anti-IL4 Rat monoclonal antibody [clone: I4.F5]
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: monoclonal
Clone: I4.F5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR