Anti-BNP Mouse monoclonal antibody unconjugated [clone: 50E1]

Proveedor: Abcam
ABCAAB20984-100EA 786 EUR
ABCAAB20984-100
Anti-BNP Mouse monoclonal antibody unconjugated [clone: 50E1]
Anticuerpos
Type: Primary
Antigen: BNP (Brain Natriuretic Peptide)
Clonality: monoclonal
Clone: 500
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR