Anti-Mumps nucleoprotein Mouse monoclonal antibody [clone: 6001]

ABCAAB20859-50EA 588 EUR
ABCAAB20859-50
Anti-Mumps nucleoprotein Mouse monoclonal antibody [clone: 6001]
Type: Primary
Antigen: Mumps nucleoprotein
Clonality: monoclonal
Clone: 6001
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR