Anti-Influenza A Virus Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB20841-100EA 0 EUR
ABCAAB20841-100
Anti-Influenza A Virus Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Influenza A Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR