Anti-Respiratory Syncytial Virus Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Abcam
ABCAAB20686-100EA 786 EUR
ABCAAB20686-100
Anti-Respiratory Syncytial Virus Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Primary
Antigen: Respiratory Syncytial Virus
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR