Anti-Islet 1 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB20670-100EA 620 EUR
ABCAAB20670-100
Anti-Islet 1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Islet 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now