Anti-LYZ Rabbit polyclonal antibody Biotin

ABCAAB20596-1EA 577 EUR
ABCAAB20596-1
Anti-LYZ Rabbit polyclonal antibody Biotin
Type: Primary
Antigen: LYZ (lysozyme)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR