Anti-PFKM Goat polyclonal antibody HRP (Horseradish Peroxidase)

ABCAAB20563-100EA 611 EUR
ABCAAB20563-100
Anti-PFKM Goat polyclonal antibody HRP (Horseradish Peroxidase)
Type: Primary
Antigen: PFKM (phosphofructokinase, muscle)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR