Anti-Gram Negative Endotoxin Mouse monoclonal antibody [clone: 301]

ABCAAB20518-200EA 604 EUR
ABCAAB20518-200
Anti-Gram Negative Endotoxin Mouse monoclonal antibody [clone: 301]
Type: Primary
Antigen: Gram Negative Endotoxin
Clonality: monoclonal
Clone: 301
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR