Anti-BCL2L1 Rabbit polyclonal antibody Alexa Fluor® 488

ABCAAB20486-100EA 588 EUR
ABCAAB20486-100
Anti-BCL2L1 Rabbit polyclonal antibody Alexa Fluor® 488
Type: Primary
Antigen: BCL2L1 (BCL2-like 1)
Clonality: polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR