Anti-COX IV Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB20393-100EA 753 EUR
ABCAAB20393-100
Anti-COX IV Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: COX IV
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR