Anti-Respiratory Syncytial Virus fusion protein Mouse monoclonal antibody [clone: BDI064]

ABCAAB20339-50EA 604 EUR
ABCAAB20339-50
Anti-Respiratory Syncytial Virus fusion protein Mouse monoclonal antibody [clone: BDI064]
Type: Primary
Antigen: Respiratory Syncytial Virus fusion protein
Clonality: monoclonal
Clone: BDI064
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR