Anti-SCG2 Mouse Monoclonal Antibody [clone: SGII 6B1/3]

Proveedor: Abcam
ABCAAB20245-50EA 0 EUR
ABCAAB20245-50
Anti-SCG2 Mouse Monoclonal Antibody [clone: SGII 6B1/3]
Anticuerpos
Type: Primary
Antigen: SCG2 (secretogranin II)
Clonality: monoclonal
Clone: SGII 6B1/3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR