Anti-RAD51D Mouse monoclonal antibody [clone: Rad51D 5B3/6]

ABCAAB20243-50EA 604 EUR
ABCAAB20243-50
Anti-RAD51D Mouse monoclonal antibody [clone: Rad51D 5B3/6]
Type: Primary
Antigen: RAD51D (RAD51 paralog D)
Clonality: monoclonal
Clone: Rad51D 5B3/6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR