Anti-TP53 Mouse monoclonal antibody [clone: PAb 1104]

ABCAAB20226-50EA 611 EUR
ABCAAB20226-50
Anti-TP53 Mouse monoclonal antibody [clone: PAb 1104]
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone: PAb 1104
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR