Anti-Maltose Binding Protein Mouse Monoclonal Antibody [clone: R3.2]

Proveedor: Abcam
ABCAAB20215-50EA 739 EUR
ABCAAB20215-50
Anti-Maltose Binding Protein Mouse Monoclonal Antibody [clone: R3.2]
Anticuerpos
Type: Primary
Antigen: Maltose Binding Protein
Clonality: monoclonal
Clone: R3.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR