Anti-KRT7 Mouse Monoclonal Antibody [clone: LP1K]

Proveedor: Abcam
ABCAAB20206-50EA 756 EUR
ABCAAB20206-50
Anti-KRT7 Mouse Monoclonal Antibody [clone: LP1K]
Anticuerpos
Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: monoclonal
Clone: LP1K
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR