Anti-EPCAM Mouse Monoclonal Antibody [clone: AUA1]
Proveedor: Abcam
ABCAAB20160-100EA 475 EUR
ABCAAB20160-100
Anti-EPCAM Mouse Monoclonal Antibody [clone: AUA1]
Anticuerpos
Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: monoclonal
Clone: AUA1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human