Anti-CDC6 Mouse Monoclonal Antibody [clone: Cdc6 9H8/5]

Proveedor: Abcam
ABCAAB20150-50EA 0 EUR
ABCAAB20150-50
Anti-CDC6 Mouse Monoclonal Antibody [clone: Cdc6 9H8/5]
Anticuerpos
Type: Primary
Antigen: CDC6 (cell division cycle 6)
Clonality: monoclonal
Clone: Cdc6 9H8/5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR