Anti-ICAM1 Mouse monoclonal antibody [clone: P1W16]

ABCAAB20144-50EA 651 EUR
ABCAAB20144-50
Anti-ICAM1 Mouse monoclonal antibody [clone: P1W16]
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone: P1W16
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR