Anti-Phenytoin Mouse monoclonal antibody [clone: 304]

ABCAAB19966-100EA 604 EUR
ABCAAB19966-100
Anti-Phenytoin Mouse monoclonal antibody [clone: 304]
Type: Primary
Antigen: Phenytoin
Clonality: monoclonal
Clone: 304
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR