Anti-SCD1 Mouse Monoclonal Antibody [clone: CD.E10]

Proveedor: Abcam
ABCAAB19862-100EA 820 EUR
ABCAAB19862-100
Anti-SCD1 Mouse Monoclonal Antibody [clone: CD.E10]
Anticuerpos
Type: Primary
Antigen: SCD1
Clonality: monoclonal
Clone: CD.E10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR