Anti-COL2 Goat polyclonal antibody Biotin

ABCAAB19752-50EA 611 EUR
ABCAAB19752-50
Anti-COL2 Goat polyclonal antibody Biotin
Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR