Anti-CD8A Mouse monoclonal antibody [clone: LT8]

ABCAAB19718-500EA 577 EUR
ABCAAB19718-500
Anti-CD8A Mouse monoclonal antibody [clone: LT8]
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone: LT8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR